Learn More
ARPC3 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305415A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p21 subunit, has yet to be determined.Specifications
ARPC3 | |
Polyclonal | |
Unconjugated | |
ARPC3 | |
actin related protein 2/3 complex subunit 3, 21kDa, actin related protein 2/3 complex, subunit 3 (21 kD), actin related protein 2/3 complex, subunit 3, 21kDa, ARC21, arp2/3 complex 21 kDa subunit, ARP2/3 protein complex subunit p21, p21-Arc | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10094, 56378 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O15145, Q9JM76 | |
Arpc3 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.