Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARPC4/p20-ARC Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982312
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2312 Supplier Bioss Supplier No. BS12528R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes one of seven subunits of the human Arp2/3 protein complex. This complex controls actin polymerization in cells and has been conserved throughout eukaryotic evolution. This gene encodes the p20 subunit, which is necessary for actin nucleation and high-affinity binding to F-actin. Alternative splicing results in multiple transcript variants. Naturally occurring read-through transcription exists between this gene and the downstream tubulin tyrosine ligase-like family, member 3 , which results in the production of a fusion protein.

Specifications

Antigen ARPC4/p20-ARC
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ARPC4
Gene Accession No. P59998
Gene Alias 20kDa; 5330419I20Rik; actin related protein 2/3 complex subunit 4; actin related protein 2/3 complex subunit 4, 20kDa; actin related protein 2/3 complex, subunit 4; actin related protein 2/3 complex, subunit 4, 20kDa; Actin-related protein 2/3 complex subunit 4; AI327076; Arc20; Arp2/3 complex 20 kDa subunit; arp2/3 protein complex subunit p20; ARPC4; BOS_20735; fa07b12; MGC13544; p20-ARC; wu:fa07b12; zgc:77932
Gene Symbols ARPC4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ARPC4/p20-ARC.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10093
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.