Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARPC4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550552

Catalog No. PA550552


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total ARPC4 protein.

This gene encodes one of seven subunits of the human Arp2/3 protein complex. This complex controls actin polymerization in cells and has been conserved throughout eukaryotic evolution. This gene encodes the p20 subunit, which is necessary for actin nucleation and high-affinity binding to F-actin. Alternative splicing results in multiple transcript variants. Naturally occurring read-through transcription exists between this gene and the downstream tubulin tyrosine ligase-like family, member 3, which results in the production of a fusion protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ARPC4
Polyclonal
Unconjugated
ARPC4
20kDa; 5330419I20Rik; actin related protein 2/3 complex subunit 4; actin related protein 2/3 complex subunit 4, 20kDa; actin related protein 2/3 complex, subunit 4; actin related protein 2/3 complex, subunit 4, 20kDa; actin-related protein 2/3 complex subunit 4; AI327076; ARC20; arp2/3 complex 20 kDa subunit; arp2/3 protein complex subunit p20; ARPC4; fa07b12; MGC13544; p20-ARC; wu:fa07b12; zgc:77932
Rabbit
Antigen affinity chromatography
RUO
10093, 68089
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P59998, P59999
ARPC4
Synthetic peptide corresponding to a region derived from internal residues of human actin related protein 2/3 complex, subunit 4, 20kDa.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.