Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARPC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568581 Supplier Invitrogen™ Supplier No. PA568581
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 77%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 90%; Zebrafish: 100%.

This gene encodes one of seven subunits of the human Arp2/3 protein complex. This complex controls actin polymerization in cells and has been conserved throughout eukaryotic evolution. This gene encodes the p20 subunit, which is necessary for actin nucleation and high-affinity binding to F-actin. Alternative splicing results in multiple transcript variants. Naturally occurring read-through transcription exists between this gene and the downstream tubulin tyrosine ligase-like family, member 3, which results in the production of a fusion protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARPC4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ARPC4
Gene Accession No. P59999
Gene Alias 20kDa; 5330419I20Rik; actin related protein 2/3 complex subunit 4; actin related protein 2/3 complex subunit 4, 20kDa; actin related protein 2/3 complex, subunit 4; actin related protein 2/3 complex, subunit 4, 20kDa; actin-related protein 2/3 complex subunit 4; AI327076; ARC20; arp2/3 complex 20 kDa subunit; arp2/3 protein complex subunit p20; ARPC4; fa07b12; MGC13544; p20-ARC; wu:fa07b12; zgc:77932
Gene Symbols ARPC4
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse ARPC4 (aa 11-60).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68089
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.