Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARPC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118927
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118927 200 μL
PIPA5122228 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5118927 Supplier Invitrogen™ Supplier No. PA5118927
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARPC4 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes one of seven subunits of the human Arp2/3 protein complex. This complex controls actin polymerization in cells and has been conserved throughout eukaryotic evolution. This gene encodes the p20 subunit, which is necessary for actin nucleation and high-affinity binding to F-actin. Alternative splicing results in multiple transcript variants. Naturally occurring read-through transcription exists between this gene and the downstream tubulin tyrosine ligase-like family, member 3, which results in the production of a fusion protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARPC4
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ARPC4
Gene Accession No. P59998, P59999
Gene Alias 20kDa; 5330419I20Rik; actin related protein 2/3 complex subunit 4; actin related protein 2/3 complex subunit 4, 20kDa; actin related protein 2/3 complex, subunit 4; actin related protein 2/3 complex, subunit 4, 20kDa; actin-related protein 2/3 complex subunit 4; AI327076; ARC20; arp2/3 complex 20 kDa subunit; arp2/3 protein complex subunit p20; ARPC4; fa07b12; MGC13544; p20-ARC; wu:fa07b12; zgc:77932
Gene Symbols ARPC4
Host Species Rabbit
Immunogen Recombinant protein Actin Related Protein 2/3 Complex Subunit 4. The antigen corresponds to amino acid range 3-165 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10093, 68089
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.