Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARPC5 Monoclonal Antibody (OTI2G1)
GREENER_CHOICE

Catalog No. PIMA526193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526193 Supplier Invitrogen™ Supplier No. MA526193
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ARPC5 Monoclonal Antibody for IHC (P), Western Blot

This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARPC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2G1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARPC5
Gene Accession No. O15511
Gene Alias 5830443F10Rik; actin related protein 2/3 complex subunit 5; actin related protein 2/3 complex subunit 5, 16kDa; actin related protein 2/3 complex, subunit 5; actin related protein 2/3 complex, subunit 5, 16kDa; actin-related protein 2/3 complex subunit 5; actin-related protein 2/3 complex subunit 5-like protein; ARC16; Arp2/3 complex 16 kDa subunit; Arp2/3 protein complex subunit p16; Arpc5; dJ127C7.3; p16-ARC
Gene Symbols ARPC5
Host Species Mouse
Immunogen Full length human recombinant protein of ARPC5 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10092
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.