Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARPC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530352
Description
Recommended positive controls: THP-1, HL-60. Predicted reactivity: Mouse (96%), Rat (97%), Xenopus laevis (82%), Pig (97%), Chicken (94%), Chimpanzee (97%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined.
Specifications
| ARPC5 | |
| Polyclonal | |
| Unconjugated | |
| ARPC5 | |
| 5830443F10Rik; actin related protein 2/3 complex subunit 5; actin related protein 2/3 complex subunit 5, 16kDa; actin related protein 2/3 complex, subunit 5; actin related protein 2/3 complex, subunit 5, 16kDa; actin-related protein 2/3 complex subunit 5; actin-related protein 2/3 complex subunit 5-like protein; ARC16; Arp2/3 complex 16 kDa subunit; Arp2/3 protein complex subunit p16; Arpc5; dJ127C7.3; p16-ARC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10092 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.99 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O15511 | |
| ARPC5 | |
| Recombinant protein encompassing a sequence within the center region of human p16 ARC. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction