Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARPC5L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554551 Supplier Invitrogen™ Supplier No. PA554551
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NFKSSEIEQA VQSLDRNGVD LLMKYIYKGF EKPTENSSAV LLQWHEKALA VGGLGSIIRV LTARKTV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

May function as component of the Arp2/3 complex which is involved in regulation of actin polymerization and together with an activating nucleation-promoting factor (NPF) mediates the formation of branched actin networks.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARPC5L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARPC5L
Gene Accession No. Q9BPX5
Gene Alias 2010015J01Rik; actin related protein 2/3 complex subunit 5 like; actin related protein 2/3 complex subunit 5-like; actin related protein 2/3 complex, subunit 5-like; actin-related protein 2/3 complex subunit 5-like protein; AI852867; ARC16-2; arp2/3 complex 16 kDa subunit 2; Arpc5l; AW555592; AW742746
Gene Symbols ARPC5L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARPC5L. Recombinant protein control fragment (Product #RP-92980).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81873
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.