Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARPM1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982313
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2313 Supplier Bioss Supplier No. BS12529R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACTRT3 is a protein coding gene.

Specifications

Antigen ARPM1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ACTRT3
Gene Accession No. Q9BYD9
Gene Alias 1700119I24Rik; actin related protein M1; actin related protein T3; actin, gamma 1 propeptide; Actin-related protein M1; actin-related protein M1 homolog; Actin-related protein T3; Actrt3; ARPM1; ARP-T3; LOC100343735; RGD1561457
Gene Symbols ACTRT3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ARPM1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84517
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.