Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARPP19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103696 Supplier Invitrogen™ Supplier No. PA5103696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARPP19.

Protein phosphatase inhibitor that specifically inhibits protein phosphatase 2A (PP2A) during mitosis. When phosphorylated at Ser-62 during mitosis, specifically interacts with PPP2R2D (PR55-delta) and inhibits its activity, leading to inactivation of PP2A, an essential condition to keep cyclin-B1-CDK1 activity high during M phase. May indirectly enhance GAP-43 expression by binding to the NGF-regulatory region of its mRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARPP19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARPP19
Gene Accession No. P56211, P56212, Q712U5
Gene Alias 19kDa; 2700024H10Rik; Arpp12; ARPP16; ARPP-16; Arpp19; ARPP-19; AW559096; cAMP regulated phosphoprotein 19; cAMP regulated phosphoprotein 19kDa; cAMP-regulated phosphoprotein 19; cAMP-regulated phosphoprotein 19kDa; cAMP-regulated phosphoprotein ARPP-19; cAMP-regulated phosphoprotein, 19kDa; cyclic AMP phosphoprotein, 19 kD; cyclic AMP phosphoprotein, 19 kDa; cyclic AMP phosphoprotein, 19kD; ENSAL
Gene Symbols ARPP19
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARPP19(Accession P56211), corresponding to amino acid residues M1-K42.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10776, 59046, 60336
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.