Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arprinocid 95.0+%, TCI America™
SDP

Catalog No. A3146250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3146250MG 250 mg
A314650MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3146250MG Supplier TCI America Supplier No. A3146250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55779-18-5
Molecular Formula C12H9ClFN5
Molecular Weight (g/mol) 277.69
MDL Number MFCD00867210
InChI Key NAPNOSFRRMHNBJ-UHFFFAOYSA-N
Synonym 9-(2-Chloro-6-fluorobenzyl)-9H-purin-6-amine, Arpocox
PubChem CID 41574
IUPAC Name 9-[(2-chloro-6-fluorophenyl)methyl]-9H-purin-6-amine
SMILES NC1=C2N=CN(CC3=C(F)C=CC=C3Cl)C2=NC=N1

Specifications

Melting Point 245°C
Quantity 250 mg
Formula Weight 277.69
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Arprinocid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.