Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584584
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584584 Supplier Invitrogen™ Supplier No. PA584584
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDLYVQTLQV VPAESSSPQG PLTVLQERLL HKLGDNAYPF TLQMVTNLPC SVT Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 85%.

May play a role in an as yet undefined retina-specific signal transduction. Could binds to photoactivated-phosphorylated red/green opsins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Arr3
Gene Accession No. P36575
Gene Alias ARR3; Arr4; arrestin 3; arrestin 3 retinal (X-arrestin); arrestin 3, retinal; arrestin 3, retinal (X-arrestin); arrestin 4; Arrestin C; ArrestinC; arrestin-C; ARRX; Car; Carfl; cArr; C-arrestin; Cone arrestin; retinal cone arrestin-3; X-arrestin
Gene Symbols Arr3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARR3. Recombinant protein control fragment (Product #RP-108483).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 407
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.