Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588911 Supplier Invitrogen™ Supplier No. PA588911
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSKVFKKTSS NGKLSIYLGK RDFVDHVDTV EPIDGVVLVD PEYLKCRKLF VMLTCAFRYG RDDLEVIGLT FRKDLYVQTL QVVPAESSSP QGPLTVLQER LLHKLGDNAY PFTLQMVTNL PCSVTLQPGP EDAGKPCGID FEVKSFCAEN PEETVSKRDY VRLVVRKVQF APPEAGPGPS AQTIRRFLLS AQPLQLQAWM DREVHYHGEP ISVNVSINNC TNKVIKKIKI SVDQITDVVL YSLDKYTKTV FIQEFTETVA; Positive Samples: Mouse eye, Rat eye.

May play a role in an as yet undefined retina-specific signal transduction. Could binds to photoactivated-phosphorylated red/green opsins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARR3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Arr3
Gene Accession No. P36576, Q9EQP6
Gene Alias ARR3; Arr4; arrestin 3; arrestin 3 retinal (X-arrestin); arrestin 3, retinal; arrestin 3, retinal (X-arrestin); arrestin 4; Arrestin C; ArrestinC; arrestin-C; ARRX; Car; Carfl; cArr; C-arrestin; Cone arrestin; retinal cone arrestin-3; X-arrestin
Gene Symbols Arr3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170735, 171107
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.