Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARRDC1-AS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144189
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144189 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144189 Supplier Invitrogen™ Supplier No. PA5144189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARRDC1-AS1 Polyclonal Antibody for IHC (P), ELISA

ARRDC1-AS1 is a gene that is also known as Arrestin Domain Containing 1 Antisense RNA 1. It is categorized as an antisense RNA, which is a type of RNA molecule that is transcribed from the opposite strand of a protein-coding gene. ARRDC1-AS1 is specifically transcribed from the antisense strand of the ARRDC1 gene.The specific function of ARRDC1-AS1 is not yet fully understood, and further research is required to elucidate its role in cellular processes and its potential implications for human health and disease. Antisense RNAs have been found to play important regulatory roles in gene expression, including the regulation of transcription, mRNA stability, and translation. Therefore, understanding the specific functions and mechanisms of ARRDC1-AS1 may provide valuable insights into the molecular processes underlying gene regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARRDC1-AS1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ARRDC1-AS1
Gene Accession No. Q9H2J1
Gene Alias C9orf37
Gene Symbols ARRDC1-AS1
Host Species Rabbit
Immunogen Recombinant Human Uncharacterized protein ARRDC1-AS1 protein (1-176AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85026
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.