Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARRDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560681 Supplier Invitrogen™ Supplier No. PA560681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IHTPRFSKDH KCSLVFYILS PLNLNSIPDI EQPNVASATK KFSYKLVKTG SVVLTASTDL RGYVVGQALQ LHADVE Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

Functions as an adapter recruiting ubiquitin-protein ligases to their specific substrates (PubMed:23886940, PubMed:27462458). Through an ubiquitination-dependent mechanism plays for instance a role in the incorporation of SLC11A2 into extracellular vesicles (PubMed:27462458). More generally, plays a role in the extracellular transport of proteins between cells through the release in the extracellular space of microvesicles (PubMed:22315426). By participating in the ITCH-mediated ubiquitination and subsequent degradation of NOTCH1, negatively regulates the NOTCH signaling pathway (PubMed:23886940). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARRDC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARRDC1
Gene Accession No. Q8N5I2
Gene Alias AI957342; alpha-arrestin 1; Arrdc1; arrestin domain containing 1; arrestin domain-containing protein 1; BC004091
Gene Symbols ARRDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARRDC1. Recombinant protein control fragment (Product #RP-93114).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92714
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.