Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARRDC2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB135012 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135012 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135012 Supplier Novus Biologicals Supplier No. NBP306285AF594
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARRDC2 Polyclonal specifically detects ARRDC2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ARRDC2
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias arrestin domain containing 2, arrestin domain-containing protein 2, CLONE24945, PP2703
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ARRDC2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27106
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.