Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARRDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599590 Supplier Invitrogen™ Supplier No. PA599590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARRDC2.

ARRDC2 gene ontology (GO) annotation includes plasma membrane and plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARRDC2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARRDC2
Gene Accession No. Q8TBH0
Gene Alias 4632416I05Rik; ARRDC2; arrestin domain containing 2; arrestin domain-containing protein 2; CLONE24945; ILAD1; induced by lysergic acid diethylamide-1; PP2703
Gene Symbols ARRDC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARRDC2(Accession Q8TBH0), corresponding to amino acid residues P372-C407.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27106
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.