Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARRDC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106544 Supplier Invitrogen™ Supplier No. PA5106544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ARRDC3.

Adapter protein that plays a role in regulating cell-surface expression of adrenergic receptors and probably also other G protein-coupled receptors. Plays a role in NEDD4-mediated ubiquitination and endocytosis of activated ADRB2 and subsequent ADRB2 degradation. May recruit NEDD4 to ADRB2. Alternatively, may function as adapter protein that does not play a major role in recruiting NEDD4 to ADRB2, but rather plays a role in a targeting ADRB2 to endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARRDC3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARRDC3
Gene Accession No. Q6TXF1, Q7TPQ9, Q96B67
Gene Alias AI450344; alpha-arrestin 3; ARRDC3; arrestin domain containing 3; arrestin domain-containing protein 3; Kiaa1376; Liver regeneration-related protein LRRG00048; LRRGT00048; mKIAA1376; TBP-2-like inducible membrane protein; thioredoxin-binding protein-2-like inducible membrane; TLIMP
Gene Symbols ARRDC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARRDC3(Accession Q96B67), corresponding to amino acid residues Q376-R414.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105171, 309945, 57561
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.