Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arrestin 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP16908320UL
Description
Arrestin 3 Polyclonal specifically detects Arrestin 3 in Human samples. It is validated for Western Blot.Specifications
| Arrestin 3 | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| arrestin 3, retinal (X-arrestin), arrestin 4, arrestin-C, ARRXcArr, CAR, C-arrestin, Cone arrestin, Retinal cone arrestin-3, X-arrestin | |
| Rabbit | |
| 43 kDa | |
| 20 μL | |
| Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Virology Bacteria and Parasites, Vision | |
| 407 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| ARR3 | |
| Synthetic peptides corresponding to ARR3 (arrestin 3, retinal (X-arrestin)) The peptide sequence was selected from the middle region of ARR3. Peptide sequence EDAGKPCGIDFEVKSFCAENPEETVSKRDYVRLVVRKVQFAPPEAGPGPS. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction