Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560148
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560148 Supplier Invitrogen™ Supplier No. PA560148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TCSLFMRNIA PNISRAEIIS LCKRYPGFMR VALSEPQPER RFFRRGWVTF DRSVNIKEIC WNLQNIRLRE CELSPGVNRD LTRRVRNING ITQHKQI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 43%.

Acts as a mediator between the cap-binding complex (CBC) and the primary microRNAs (miRNAs) processing machinery during cell proliferation. Contributes to the stability and delivery of capped primary miRNA transcripts to the primary miRNA processing complex containing DGCR8 and DROSHA, thereby playing a role in RNA-mediated gene silencing (RNAi) by miRNAs. Binds capped RNAs (m7GpppG-capped RNA); however interaction is probably mediated via its interaction with NCBP1/CBP80 component of the CBC complex. Involved in cell cycle progression at S phase. Does not directly confer arsenite resistance but rather modulates arsenic sensitivity. Independently of its activity on miRNAs, necessary and sufficient to promote neural stem cell self-renewal. Does so by directly binding SOX2 promoter and positively regulating its transcription. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARS2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRRT
Gene Accession No. Q9BXP5
Gene Alias 2810019G02Rik; Ars2; arsenate resistance protein 2; arsenate resistance protein ARS2; arsenite resistance protein; arsenite resistance protein 2; arsenite-resistance protein 2; Asr2; ASR2A; ASR2B; ASR2C; ASR2D; ch211-283g2.4; CHUNP6881; LOC686980; serrate; Serrate RNA effector molecule homolog; serrate RNA effector molecule homolog (Arabidopsis); serrate, RNA effector molecule; similar to arsenate resistance protein 2; srrt; zgc:92060
Gene Symbols SRRT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARS2. Recombinant protein control fragment (Product #RP-92626).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51593
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.