Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARS2 Rabbit anti-Human, Zebrafish, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572269
Description
Acts as a mediator between the cap-binding complex (CBC) and the primary microRNAs (miRNAs) processing machinery during cell proliferation. Contributes to the stability and delivery of capped primary miRNA transcripts to the primary miRNA processing complex, thereby playing a role in RNA-mediated gene silencing (RNAi) by miRNAs (By similarity). Independently of its activity on miRNAs, necessary and sufficient to promote neural stem cell self-renewal. Does so by directly binding SOX2 promoter and positively regulating its transcription (By similarity).
Specifications
ARS2 | |
Polyclonal | |
Unconjugated | |
SRRT | |
Serrate RNA effector molecule homolog; Arsenite-resistance protein 2; srrt; ars2 | |
Rabbit | |
Protein A | |
RUO | |
192311, 51593 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.95 mg/mL | |
PBS with 0.09% sodium azide | |
Q66I22, Q9BXP5 | |
SRRT | |
KLH conjugated synthetic peptide between 733-766 amino acids from Zebrafish ARS2 | |
200 μL | |
Primary | |
Human, Zebrafish | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction