Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARSA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578824
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578824 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578824 Supplier Invitrogen™ Supplier No. PA578824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Testis Tissue, Rat Pancreas Tissue, Rat Skeletal Muscle Tissue, Mouse Kidney Tissue, MCF-7 whole cell. IHC: Rat Lung tissue, Human Mammary Cancer tissue. ICC/IF: U20S cell. Flow: Hela cell, PC-3 cell.

The protein encoded by this gene hydrolyzes cerebroside sulfate to cerebroside and sulfate. Defects in this gene lead to metachromatic leucodystrophy, a progressive demyelination disease which results in a variety of neurological symptoms and ultimately death. Alternatively spliced transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARSA
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene ARSA
Gene Accession No. P15289, P50428
Gene Alias ARSA; ARSA1; ARSA-I; Arylsulfatase A; Arylsulfatase A component B; Arylsulfatase A component C; As2; As-2; ASA; AS-A; ASNA-I; ATPase ASNA1; AW212749; cerebroside-sulfatase; EC 3.1.6.8; GET3; hASNA-I; MLD; TISP73; TRC40
Gene Symbols ARSA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human ARSA (454-482aa QALKQLQLLKAQLDAAVTFGPSQVARGED).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11883, 315222, 410
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.