Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Arsenal™ 5093 Quick Adjust Reflective Suspenders

Quick Adjust Suspenders are designed to be quickly attached and feature shoulder padding for comfort.

Specifications

Color Red
For Use With (Application) Firefighters, EMTs, Biochem, Hazmat
Material 600D Nylon
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-819-818
View Documents
Ergodyne™
13349
Small N/A N/A N/A
19-819-819
View Documents
Ergodyne™
13350
Medium N/A N/A N/A
19-819-821
View Documents
Ergodyne™
13351
Large N/A N/A N/A
19-819-822
View Documents
Ergodyne™
13352
X-Large N/A N/A N/A
Description

Description

  • Heavy-duty polyester webbing
  • H-shaped back design
  • Foam padded shoulder straps
  • Reflective trim through center
Specifications

Specifications

Red
Firefighters, EMTs, Biochem, Hazmat
600D Nylon
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.