Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic AA Standard, 1000 ppm As in 3% H2SO4, Ricca Chemical
SDP

Catalog No. p-3251321 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAS1KS-100 100 mL Natural LDPE Bottle
AAS1KS-500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAS1KS-100 Supplier Ricca Chemical Company Supplier No. AAS1KS100
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1327-53-3 , 7664-93-9 , 1327-53-3 , 7732-18-5
Molecular Formula As2O3
Molecular Weight (g/mol) 197.84
MDL Number MFCD00003433
InChI Key IKWTVSLWAPBBKU-UHFFFAOYSA-N
IUPAC Name diarsorosooxidane
SMILES O=[As]O[As]=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm As
CAS Min % 93.0, 5.12, 0.12
CAS Max % 97.0, 5.33, 0.13
Equivalence 1 mL = 1 mg As
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Quantity 100 mL
Chemical Name or Material Arsenic AA Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.