Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(II) sulfide, tech. 90%

Catalog No. p-7049647
Click to view available options
Quantity:
5 g
25 g

CAS: 12044-79-0 | AsH3S | 110.01 g/mol

Arsenic(II) sulfide is used as an primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12044-79-0
Molecular Formula AsH3S
Molecular Weight (g/mol) 110.01
MDL Number MFCD00049841
InChI Key BMOFEAKQXGGUPE-UHFFFAOYSA-N
Synonym arsenic ii sulfide, realgar ass, diarsanedithione
IUPAC Name arsanethiol
SMILES S[AsH2]

Specifications

Boiling Point 565°C
Melting Point ∼307°C
Physical Form Powder
Quantity 5 g
UN Number UN1557
Solubility Information Insoluble in water. Soluble in acids and alkalis.
Formula Weight 213.97
Percent Purity 90%
Grade Technical
Odor Odorless
Density 3.5 g/mL
Chemical Name or Material Arsenic(II) sulfide
Show More Show Less
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.