Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(III) chloride, Reagent Grade, Thermo Scientific Chemicals

Catalog No. aa3149930
Click to view available options
:
250g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 130.2°C
Melting Point -16°C
CAS 7784-34-1
Physical Form Liquid
Quantity 250 g
Molecular Formula AsCl3
MDL Number MFCD00014165
UN Number UN1560
Merck Index 14,801
Synonym trichloroarsine, arsenic trichloride, arsenic iii chloride, arsenic chloride, arsenous trichloride, arsenic butter, arsenous chloride, arsenious chloride, butter of arsenic, chlorure arsenieux
Solubility Information Decomposes in water with formation of As(OH)3 and HCl; Increasing dilutions in water result in As2O3 precipitation
InChI Key OEYOHULQRFXULB-UHFFFAOYSA-N
SMILES Cl[As](Cl)Cl
IUPAC Name trichloroarsane
Molecular Weight (g/mol) 181.272
PubChem CID 24570
ChEBI CHEBI:63952
Formula Weight 181.27
Grade Reagent
Odor Acrid
Sensitivity Moisture sensitive
Density 2.163 g/mL
Chemical Name or Material Arsenic(III) chloride
Show More Show Less
Hazard Category H300
Hazard Statement GHS H Statement
H300-H331-H350-H314
Fatal if swallowed.
Toxic if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ARSENIC TRICHLORIDE
EINECSNumber 232-059-5
RTECSNumber CG1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.