Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(III) iodide, 99.999% (metals basis)

Catalog No. AA4196214
Click to view available options
Quantity:
5 g
25 g

CAS: 7784-45-4 | AsI3 | 455.635 g/mol

Arsenic(III) iodide is useful for preparing organoarsenic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-45-4
Molecular Formula AsI3
Molecular Weight (g/mol) 455.635
MDL Number MFCD00014170
InChI Key IKIBSPLDJGAHPX-UHFFFAOYSA-N
Synonym arsenous triiodide, arsenic iii iodide, arsenic iodide, arsenic triiodide, triiodoarsine, arsenous iodide, hsdb 420, arsen iii-jodid
PubChem CID 24575
IUPAC Name triiodoarsane
SMILES [As](I)(I)I

Specifications

Boiling Point 424°C
Melting Point 140°C
Physical Form Powder
Quantity 25 g
UN Number UN1557
Merck Index 14,803
Solubility Information Hydrolyzes slowly in water.
Formula Weight 455.64
Percent Purity 99.999%
Odor Pungent
Sensitivity Moisture sensitive
Density 4.39 g/mL
Assay (metals basis)
Chemical Name or Material Arsenic(III) iodide
Show More Show Less
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 232-068-4
RTECSNumber CG1950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.