Learn More
Arsenic(III) oxide, 99.99% (metals basis)
Description
used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 465°C (sublimation) |
| Melting Point | 312.3°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 99.99% (Metals basis) |
| Solubility Information | Soluble in dilute acids and alkalies.Insoluble in organic solvents.Slightly soluble in water. |
| Formula Weight | 197.84 |
| Odor | Odorless |
| Density | 3.74 |
| Show More |
Safety and Handling
| Hazard Category | H300-H314-H335-H350-H371 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: II |
| EINECSNumber | 215-481-4 |
| RTECSNumber | CG3325000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.