Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenic(III) sulfide, 99.99%, Thermo Scientific™

Catalog No. ac192511000
Click to view available options
Quantity:
100 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-33-9
Molecular Formula As2S3
Molecular Weight (g/mol) 246.02
MDL Number MFCD00003435
InChI Key CUGMJFZCCDSABL-UHFFFAOYSA-N
IUPAC Name diarsenic(3+) trisulfanediide
SMILES [S--].[S--].[S--].[As+3].[As+3]

Specifications

Color Yellow to Orange
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99%
Linear Formula AS2S3
Merck Index 15, 797
Formula Weight 246.02
Chemical Name or Material Arsenic(III) sulfide, 99.99%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.