Learn More
Arsenic Trioxide, Powder, ACS, 99.95-100.05%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 125 g |
| Percent Purity | 99.95 to 100.05% |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
| Residue after Ignition | 0.0002 |
| Chemical Name or Material | Arsenic Trioxide, Powder |
Safety and Handling
| Hazard Category | c |
CAUTION: For manufacturing or laboratory use only. Not for pesticide use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.