Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Arsenic(V) oxide, 99.9% (metals basis)
Click to view available options
Quantity:
25 g
100 g
Description
Arsenic(V) oxide is used in the preparation of metal arsenates. It is used for dye and printing industries. Further, it is used in wood preservatives, colored glass and metal adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 315°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.9% (metals basis) |
UN Number | UN1559 |
Merck Index | 14,798 |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 229.79 |
Percent Purity | 99.9% |
Show More |
Safety and Handling
Hazard Category | H301+H331-H350 |
Hazard Statement | GHS H Statement H301-H331-H350 Toxic if swallowed. Toxic if inhaled. May cause cancer. |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: ARSENIC PENTOXIDE |
EINECSNumber | 215-116-9 |
RTECSNumber | CG2275000 |
TSCA | Yes |
storageNote1 | Hygroscopic |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction