Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arsenious Acid Solution, For Iodide, LabChem™

Catalog No. LC115201 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC115201 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC115201 Supplier LabChem, Inc. Supplier No. LC115201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 1327-53-3 , 7664-93-9
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Color Colorless
CAS Max % 99.46, 0.5, 0.04
Linear Formula As2O3
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 197.84
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Arsenic and its oxides; Sulfur compounds
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Arsenious Acid Solution, For Iodide
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause cancer.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.