Learn More
ARSG Mouse anti-Human, Clone: OTI4H3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812732

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Sulfatases, such as ARSG, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules. This gene, a novel arylsulfatase, has activity toward pseudosubstrates including p-nitrocatechol sulfate and 4-methylumbelliferyl sulfate. Activity is competitively inhibited by phosphate. The unprocessed protein is active as a 63-kDa monomer and demonstrates an acidic pH optimum as typically seen for lysosomal sulfatases. The protein accumulates within lysosomes and is also a glycoprotein that binds specifically to mannose 6-phosphate receptors.Specifications
ARSG | |
Monoclonal | |
Unconjugated | |
ARSG | |
arylsulfatase G | |
Mouse | |
Affinity Chromatography | |
RUO | |
22901 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96EG1 | |
ARSG | |
Human recombit protein fragment corresponding to amino acids 17-525 of human ARSG produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.