Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARSI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558375
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558375 Supplier Invitrogen™ Supplier No. PA558375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SADPYEREDL AGQRPDVVRT LLARLAEYNR TAIPVRYPAE NPRAHPDFNG GAWGPWASDE EEE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

Sulfatases (EC 3.1.5.6), such as ARSI, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARSI
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARSI
Gene Accession No. Q5FYB1
Gene Alias Arsi; arylsulfatase family member I; arylsulfatase family, member I; arylsulfatase I; ASI; BC020108; RGD1310242; SPG66
Gene Symbols ARSI
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARSI. Recombinant protein control fragment (Product #RP-103855).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340075
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.