Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARSJ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557678
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557678 Supplier Invitrogen™ Supplier No. PA557678
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TADPYERVDL SNRYPGIVKK LLRRLSQFNK TAVPVRYPPK DPRSNPRLNG GVWGPWYKEE TKK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 52%.

ARSJ is a sulfatase that hydrolyzes sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. Sulfatases are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARSJ
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARSJ
Gene Accession No. Q5FYB0
Gene Alias 9330196J05Rik; Arsj; arylsulfatase family member J; arylsulfatase family, member J; arylsulfatase J; ASJ; D830047F08; RGD1307640; UNQ372/PRO708
Gene Symbols ARSJ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARSJ. Recombinant protein control fragment (Product #RP-104163).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79642
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.