Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARSJ Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103619
Description
Antibody detects endogenous levels of total ARSJ.
ARSJ is a sulfatase that hydrolyzes sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. Sulfatases are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules.
Specifications
ARSJ | |
Polyclonal | |
Unconjugated | |
ARSJ | |
9330196J05Rik; Arsj; arylsulfatase family member J; arylsulfatase family, member J; arylsulfatase J; ASJ; D830047F08; RGD1307640; UNQ372/PRO708 | |
Rabbit | |
Affinity chromatography | |
RUO | |
271970, 311013, 79642 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5FYB0, Q8BM89 | |
ARSJ | |
A synthesized peptide derived from human ARSJ(Accession Q5FYB0), corresponding to amino acid residues C371-N421. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction