Learn More
Invitrogen™ ARSK Polyclonal Antibody

Description
Immunogen sequence: WSPLSEMHPV DYYSSYTKNC TGRFTKKEIK NIRAFYYAMC AETDAMLGEI ILALHQLDLL QKTIVIYSSD HGELAMEHRQ FYKMSMYEAS AHVPLLMMGP GIKAGLQVSN VVSLVDIYPT MLDIAGIPLP QNLSGYSLLP LSSETFKNE Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
Specifications
Specifications
| Antigen | ARSK |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ARSK |
| Gene Accession No. | Q6UWY0 |
| Gene Alias | 2810429K17Rik; 4833414G15Rik; ARSK; arylsulfatase family member K; arylsulfatase family, member K; arylsulfatase K; ASK; RGD1310182; Telethon sulfatase; TSULF; UNQ630/PRO1246 |
| Gene Symbols | ARSK |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.