Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips PROMO

Catalog No. 02682265PR
Click to view available options
Volume (Metric):
10 μL
200 μL

Perform pipetting accurately and consistently with Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips, which seal when exposed to potential contaminants.

Features:
  • Stackable hinged racks save space in your lab
  • Sterile tips are certified free from RNase, DNase and pyrogen

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
TRUSTED_SUSTAINABILITY

Specifications

Description MicroPoint, reload insert, low retention
Volume (Metric) 10 μL
Filtered Filtered
Sterility Sterile
Model ART 10
Length (English) 1.23 in.
Length (Metric) 3.13cm
Clean Claims DNase, RNase, DNA, ATP, Bioburden, PCR Inhibitor, Endotoxin, and Pyrogen Free
Format Hinged Rack
For Use With (Equipment) Universal Fit
Product Line ART
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.