Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ART™ Barrier Pipette Tips in Bulk Packaging
GREENER_CHOICE

Catalog No. p-7146707
Click to view available options
Volume (Metric):
200 μL
5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo Scientific™ ART™ Barrier Pipette tips seal in contaminants, providing bi-directional protection for samples and pipettes. Fits most pipettes.

ART™ Barrier tips are not the same as traditional filter pipette tips. ART Barrier tips contain a one-of-a-kind, self-sealing barrier that seals in aerosol and liquid contaminants on contact, providing bi-directional protection from contamination for pipettes and samples.

  • Universal Tip, fits most brands of pipettes
  • ART™ tips are certified to be free DNase, RNase, DNA, ATP, Endotoxin, and Pyrogens
  • Convenient bulk packaging for high volume usage

  • Sterile
  • Low Retention
  • Racked with lift-off lid
TRUSTED_SUSTAINABILITY

Specifications

Description ART 5000, Filtered, Sterile, Bulk
Volume (Metric) 5 mL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Model ART 5000
Clean Claims DNase, RNase, DNA, ATP, Bioburden, PCR Inhibitor, Endotoxin, and Pyrogen Free
For Use With (Application) Cell Culture, Pipetting Large volumes of liquids where great accuracy and precision is needed
Format Bulk
Tip Style Macro
Packaging Bag of 250
Quantity 250 tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Product Line ART
Material Virgin Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.