Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ART1 Antibody (OTI6E2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270211H
Description
ART1 Monoclonal specifically detects ART1 in Human samples. It is validated for Western Blot.Specifications
ART1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ART1 | |
Recombinant protein fragment corresponding to amino acids 83-295 of human ART1 (NP_004305) produced in E.coli. | |
0.1 mL | |
417.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI6E2 | |
Western Blot | |
ADP-ribosyltransferase 1, ADP-ribosyltransferase 2, CD296, GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1, MGC133217, mono(ADP-ribosyl)transferase 1, RT6 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction