Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ART1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562332 Supplier Invitrogen™ Supplier No. PA562332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IQLDMALASF DDQYAGCAAA MTAALPDLNH TEFQANQVYA DSWTLASSQW QERQARWPEW SLSPTRPSPP PLGFRDEHGV ALLAYT Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 71%.

ADP-ribosyltransferase catalyzes the ADP-ribosylation of arginine residues in proteins. Mono-ADP-ribosylation is a posttranslational modification of proteins that is interfered with by a variety of bacterial toxins including cholera, pertussis, and heat-labile enterotoxins of E. coli. The amino acid sequence of ART1 consists of predominantly hydrophobic N- and C-terminal regions, which is characteristic of glycosylphosphatidylinositol (GPI)-anchored proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ART1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ART1
Gene Accession No. P52961
Gene Alias ADP-ribosyltransferase 1; ADP-ribosyltransferase 2; ADP-ribosyltransferase C2 and C3 toxin-like 1; ADPRT; ART1; Art2; ARTC1; CD296; GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1; mono(ADP-ribosyl)transferase 1; MonoADP-ribosyltransferase; RT6; Yac-1
Gene Symbols ART1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ART1. Recombinant protein control fragment (Product #RP-101277).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 417
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.