Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ART1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107067
Description
Sequence homology: Cow: 86%; Dog: 85%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 85%; Rat: 100%.
ADP-ribosyltransferase catalyzes the ADP-ribosylation of arginine residues in proteins. Mono-ADP-ribosylation is a posttranslational modification of proteins that is interfered with by a variety of bacterial toxins including cholera, pertussis, and heat-labile enterotoxins of E. coli. The amino acid sequence of ART1 consists of predominantly hydrophobic N- and C-terminal regions, which is characteristic of glycosylphosphatidylinositol (GPI)-anchored proteins.
Specifications
| ART1 | |
| Polyclonal | |
| Unconjugated | |
| ART1 | |
| ADP-ribosyltransferase 1; ADP-ribosyltransferase 2; ADP-ribosyltransferase C2 and C3 toxin-like 1; ADPRT; ART1; Art2; ARTC1; CD296; GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1; mono(ADP-ribosyl)transferase 1; MonoADP-ribosyltransferase; RT6; Yac-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 417 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| P52961 | |
| ART1 | |
| Synthetic peptide directed towards the C-terminal region of human ART1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction