Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ART1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298460AF647
Description
ART1 Polyclonal specifically detects ART1 in Cynomolgus Monkey samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ART1 | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ADP-ribosyltransferase 1, ADP-ribosyltransferase 2, CD296, GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1, MGC133217, mono(ADP-ribosyl)transferase 1, RT6 | |
Produced in rabbits immunized with purified, recombinant ART1 (XP_005579037.1; Gln23-Asn294). | |
0.1 mL | |
Primary | |
Cynomolgus Monkey | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
417 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction