Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ART2.2/Art2b Antibody (Nika102), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10065853AF700
Description
ART2.2/Art2b Monoclonal specifically detects ART2.2/Art2b in Mouse samples. It is validated for Flow Cytometry.Specifications
ART2.2/Art2b | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ART2B | |
pME.CD8LF-ART2.2 | |
0.125 mL | |
Primary | |
NB100-65853 recognizes the gene product of the mouse ADP-ribosyltransferase (ART) 2.2 gene, an ortholog of rat RT6. ART2.2 expressed exclusively by mature T-cells. Levels of ART2.2 expression vary between inbred strains of mice and is absent in NZW mice, in which the ART2.2 gene is deleted (PMID: 10570289, Koch-Nolte et al. 1999). | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
Nika102 | |
Flow Cytometry | |
ADP-ribosyltransferase 2b, Rt6, Rt-6, Rt6.2, Rt6-2 | |
Rat | |
Protein G purified | |
RUO | |
11872 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction