Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ART2.2 Monoclonal Antibody (Nika102)
GREENER_CHOICE

Catalog No. PIMA528162
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528162 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528162 Supplier Invitrogen™ Supplier No. MA528162
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This antibody recognizes the gene product of the mouse ADP-ribosyltransferase (ART) 2.2 gene, an ortholog of rat RT6. ART2.2 expressed exclusively by mature T-cells.

ART2.2 recognizes the gene product of the mouse ADP-ribosyltransferase (ART) 2.2 gene, an ortholog of rat RT6. ART2.2 expressed exclusively by mature T-cells. Levels of ART2.2 expression vary between inbred strains of mice and is absent in NZW mice, in which the ART2.2 gene is deleted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ART2.2
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone Nika102
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Art2b
Gene Accession No. O35975
Gene Alias ADP-ribosyltransferase 2b; ADP-ribosyltransferase C2 and C3 toxin-like 2; Art; ART2.2; Art2b; ARTC2; Ly92b; mono(ADP-ribosyl)transferase 2B; NAD(+) glycohydrolase; Rt6; Rt-6; Rt6.2; Rt6-2; T-cell differentiation marker Rt6 homolog 2; T-cell ecto-ADP-ribosyltransferase 2; T-cell mono(ADP-ribosyl)transferase 2; T-cell NAD(P)(+)--arginine ADP-ribosyltransferase 2
Gene Symbols Art2b
Host Species Rat
Immunogen pMECD8LF-ART22.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11872
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.