Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ART3 Antibody (063), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290224MFV500
Description
ART3 Monoclonal antibody specifically detects ART3 in Human samples. It is validated for ELISASpecifications
ART3 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
419 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
063 | |
ELISA | |
ADP-ribosyltransferase 3, EC 2.4.2.31, ecto-ADP-ribosyltransferase 3, FLJ26404, Mono(ADP-ribosyl)transferase 3, mono-ADP-ribosyltransferase, NAD(P)(+)--arginine ADP-ribosyltransferase 3, TMART | |
Rabbit immunized with purified, recombinant Human ART3 (Accession#: AAH08397.1, Met1-Ser362) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction