Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ART3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580354
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ART3.
Mono-ADP-ribosylation involves the transfer of the ADP-ribose moiety from NAD+ to a specific amino acid in the target protein. The rodent mono-ADP-ribosyltransferase RT6 is a glycosylphosphatidylinositol (GPI)-anchored membrane protein specifically expressed at the cell surface of rat and mouse T lymphocytes. The predicted 367-amino acid human ART3 protein has an estimated molecular mass of 41.5 kD and contains a hydrophobic peptide signal at its N terminus, 3 consensus motifs specific to enzymes catalyzing ADP-ribose transfer, a hydrophobic C-terminal sequence characteristic of a GPI-anchored protein, a novel motif repeated 3 times at its C terminus, and 1 potential glycosylation site.1 The ART3 and rodent RT6 proteins share 35% amino acid identity.
Specifications
ART3 | |
Polyclonal | |
Unconjugated | |
ART3 | |
4930569O04Rik; ADP-ribosyltransferase 3; ADP-ribosyltransferase C2 and C3 toxin-like 3; Art3; ART3 mono(ADP-ribosyl)transferase; ARTC3; Ecto-ADP-ribosyltransferase 3; mono(ADP-ribosyl)transferase 3; mono-ADP-ribosyltransferase; mono-ADP-ribosyltransferase 3; NAD(P)(+)--arginine ADP-ribosyltransferase 3; TMART | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
419 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with no preservative | |
Q13508 | |
ART3 | |
Recombinant Human ART3 Protein (Met1-Ser362). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction