Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ART3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501730580 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0580 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0580 Supplier Proteintech Group Inc Supplier No. 159301AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Mono-ADP-ribosylation involves the transfer of the ADP-ribose moiety from NAD+ to a specific amino acid in the target protein. The rodent mono-ADP-ribosyltransferase RT6 is a glycosylphosphatidylinositol (GPI)-anchored membrane protein specifically expressed at the cell surface of rat and mouse T lymphocytes. The predicted 367-amino acid human ART3 protein has an estimated molecular mass of 41.5 kD and contains a hydrophobic peptide signal at its N terminus, 3 consensus motifs specific to enzymes catalyzing ADP-ribose transfer, a hydrophobic C-terminal sequence characteristic of a GPI-anchored protein, a novel motif repeated 3 times at its C terminus, and 1 potential glycosylation site.1 The ART3 and rodent RT6 proteins share 35% amino acid identity.

Specifications

Antigen ART3
Applications Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ART3
Gene Accession No. Q13508, Q8R2G4
Gene Alias ADP ribosyltransferase 3, ART3, Ecto ADP ribosyltransferase 3, Mono(ADP ribosyl)transferase 3, TMART
Gene Symbols ART3
Host Species Rabbit
Immunogen ART3 Fusion Protein Ag8775
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109979, 419
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.