Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ART3 Recombinant Rabbit Monoclonal Antibody (063)
GREENER_CHOICE

Catalog No. PIMA530068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530068 Supplier Invitrogen™ Supplier No. MA530068
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human ART3.

Mono-ADP-ribosylation involves the transfer of the ADP-ribose moiety from NAD+ to a specific amino acid in the target protein. The rodent mono-ADP-ribosyltransferase RT6 is a glycosylphosphatidylinositol (GPI)-anchored membrane protein specifically expressed at the cell surface of rat and mouse T lymphocytes. The predicted 367-amino acid human ART3 protein has an estimated molecular mass of 41.5 kD and contains a hydrophobic peptide signal at its N terminus, 3 consensus motifs specific to enzymes catalyzing ADP-ribose transfer, a hydrophobic C-terminal sequence characteristic of a GPI-anchored protein, a novel motif repeated 3 times at its C terminus, and 1 potential glycosylation site.1 The ART3 and rodent RT6 proteins share 35% amino acid identity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ART3
Applications ELISA
Classification Recombinant Monoclonal
Clone 063
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ART3
Gene Accession No. Q13508
Gene Alias 4930569O04Rik; ADP-ribosyltransferase 3; ADP-ribosyltransferase C2 and C3 toxin-like 3; Art3; ART3 mono(ADP-ribosyl)transferase; ARTC3; Ecto-ADP-ribosyltransferase 3; mono(ADP-ribosyl)transferase 3; mono-ADP-ribosyltransferase; mono-ADP-ribosyltransferase 3; NAD(P)(+)--arginine ADP-ribosyltransferase 3; TMART
Gene Symbols ART3
Host Species Rabbit
Immunogen Recombinant Human ART3 Protein (Met1-Ser362).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 419
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.