Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ART4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | ART4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
ART4 Polyclonal specifically detects ART4 in Human samples. It is validated for Western Blot.Specifications
ART4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
420 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
ADP-ribosyltransferase 4 (DO blood group), ADP-ribosyltransferase 4 (Dombrock blood group), CD297, CD297 antigen, DODombrock blood group, DOK1ADP-ribosyltransferase 4, Dombrock blood group carrier molecule, EC 2.4.2.31, ecto-ADP-ribosyltransferase 4, Mono(ADP-ribosyl)transferase 4, mono-ADP-ribosyltransferase 4, NAD(P)(+)--arginine ADP-ribosyltransferase 4 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human ART4 (NP_066549.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title